Ethylene Glycol

Catalog number:  29810B

Thermo Scientific™  Related applications: Antibody Labeling | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Ethylene Glycol Solution provides exceptional antifreeze protection and storage stability for antibody-enzyme conjugates because it is purified to remove impurities commonly found traditional glycerol stocks.

Features of Ethylene Glycol:

• Specially purified to remove impurities such as aldehydes, peroxides, iron and UV absorbing hydrocarbons
• Suitable for enzyme storage without the worry of losing enzymatic activity
• Stable for months

This product is a 50% (w/v) aqueous solution of highly purified ethylene glycol. When mixed in equal volume with purified protein samples, such as primary antibodies, the solution stabilizes and maintains the mixture as a liquid during freezer storage (-20°). Ethylene glycol is a suitable alternative to glycerol for most protein-storage applications. In fact, this preparation of ethylene glycol is devoid of various common glycerol impurities and potiential degradation products that are damaging to certain enzymes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Size: Custom
Reagent Type: Stabilizing Reagent
Validated Application: ELISA, Immunohistochemistry, Western Blot

Contents & storage

Store at room temperature.

Documents

Manuals & protocols