4-Methyl-2-pentenal, 95%
4-Methyl-2-pentenal, 95%
4-Methyl-2-pentenal, 95%
Thermo Scientific Chemicals

4-Methyl-2-pentenal, 95%

CAS: 5362-56-1 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 298350250
Price (USD)
168.65
Online exclusive
187.00
Save 18.35
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
168.65
Online exclusive
187.00
Save 18.35
Each
Chemical Identifiers
CAS5362-56-1
IUPAC Name(2Z)-4-methylpent-2-enal
Molecular FormulaC6H10O
InChI KeyRIWPMNBTULNXOH-ARJAWSKDSA-N
SMILESCC(C)\C=C/C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4360 to 1.4440 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=94 %
Acid value=<15 mg KOH/g
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only