Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%
Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%
Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%
Thermo Scientific Chemicals

Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%

CAS: 142547-15-7 | C8H16ClNO2 | 193.67 g/mol
Have Questions?
Catalog NumberQuantity
298392500250 mg
Catalog number 298392500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS142547-15-7
IUPAC Nameethyl 2-aminocyclopentane-1-carboxylate hydrochloride
Molecular FormulaC8H16ClNO2
InChI KeyRYBMXCZWUCHLJB-UHFFFAOYNA-N
SMILESCl.CCOC(=O)C1CCCC1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Melting point97°C to 100°C
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only