Ethyl isocyanoacetate, 97%
Ethyl isocyanoacetate, 97%
Ethyl isocyanoacetate, 97%
Thermo Scientific Chemicals

Ethyl isocyanoacetate, 97%

CAS: 2999-46-4 | C5H7NO2 | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 299551000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2999-46-4
IUPAC Nameethyl 2-isocyanoacetate
Molecular FormulaC5H7NO2
InChI KeyFPULFENIJDPZBX-UHFFFAOYSA-N
SMILESCCOC(=O)C[N+]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4170 to 1.4250 (20°C, 589 nm)
GC>=96.0 %
Appearance (Color)Clear amber to dark brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only