N-Iodosuccinimide, 98%
N-Iodosuccinimide, 98%
N-Iodosuccinimide, 98%
Thermo Scientific Chemicals

N-Iodosuccinimide, 98%

CAS: 516-12-1 | C4H4INO2 | 224.99 g/mol
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 299575000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS516-12-1
IUPAC Name1-iodopyrrolidine-2,5-dione
Molecular FormulaC4H4INO2
InChI KeyLQZMLBORDGWNPD-UHFFFAOYSA-N
SMILESIN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Crystalline powder
Melting point197°C to 204°C
Titration Iodometric>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only