Regulatory requirements vary by country; products may not be available in your geographic area.

Prionics®-Check PrioSTRIP BSE Kit

Catalog number: 30000

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Prionics®-Check PrioSTRIP BSE Kit is an immunochromatographic assay based on the antibody-mediated detection of the protease-resistant form of the prion protein, PrPSc. The unique 8-strip PrioSTRIP® cassettes are simply dipped into the homogenized and digested brain-detection antibody mixture that then runs up the strips until captured by an immobilized antibody. The result is clear-cut: two lines means BSE positive; one line mean BSE negative.
For Veterinary Use Only.

Specifications

Target Organism Class: Prions
Validated Application: Immunochromatographic Test
Detection Method: Immunochromatographic
Disease: Bovine Spongiform Encephalopathy (BSE)
Product Line: PrioSTRIP™, Prionics®-Check, Prionics™
Product Size: 470 samples
Species: Cattle⁄Bovine

Documents

Manuals & protocols