Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Thermo Scientific Chemicals

Tetrakis(1-pyrazolyl)borate, potassium salt, 95%

CAS: 14782-58-2 | C12H12BKN8 | 318.2 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 300160010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14782-58-2
IUPAC Namepotassium tetrakis(1H-pyrazol-1-yloxy)boranuide
Molecular FormulaC12H12BKN8O4
InChI KeyRKBGLZQVHBLUBM-UHFFFAOYSA-N
SMILES[K+].O(N1C=CC=N1)[B-](ON1C=CC=N1)(ON1C=CC=N1)ON1C=CC=N1
さらに表示
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only