2-Amino-6-chlorobenzoic acid, 99%
2-Amino-6-chlorobenzoic acid, 99%
2-Amino-6-chlorobenzoic acid, 99%
Thermo Scientific Chemicals

2-Amino-6-chlorobenzoic acid, 99%

CAS: 2148-56-3 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 300500100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2148-56-3
IUPAC Name2-amino-6-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeySZCPTRGBOVXVCA-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point143°C to 147°C
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Light yellow to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only