Lithium isopropoxide, pure
Lithium isopropoxide, pure
Lithium isopropoxide, pure
Thermo Scientific Chemicals

Lithium isopropoxide, pure

CAS: 2388-10-5 | C3H7LiO | 66.03 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 301240250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2388-10-5
IUPAC Namelithium(1+) propan-2-olate
Molecular FormulaC3H7LiO
InChI KeyHAUKUGBTJXWQMF-UHFFFAOYSA-N
SMILES[Li+].CC(C)[O-]
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to yellow to beige to tan
Infrared spectrumConforms
Appearance (Form)Powder
Titration>=94 % (corrected for LiOH+Li2CO3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only