1-Octene, 99+%
1-Octene, 99+%
1-Octene, 99+%
Thermo Scientific Chemicals

1-Octene, 99+%

CAS: 111-66-0 | C8H16 | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 301250025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-66-0
IUPAC Nameoct-1-ene
Molecular FormulaC8H16
InChI KeyKWKAKUADMBZCLK-UHFFFAOYSA-N
SMILESCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 %
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only