Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%
Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%
Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%
Acros Organics

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%

CAS: 65501-24-8 | C10H13K3N2O8·2H2O | 442.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 301601000
also known as 30160
HSN 29224990
Price (INR)
14,321.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS65501-24-8
IUPAC Nametripotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H17K3N2O10
InChI KeyMAPFUJCWRWFQIY-UHFFFAOYSA-K
SMILESO.O.[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.0 % (on dry substance)
Appearance (Color)White
Water=<9 % (K.F.)
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only