Hypersil™ MOS C8 HPLC Columns
Thermo Scientific™

Hypersil™ MOS C8 HPLC Columns

Use as a general-purpose C8 phase for acidic and neutral analytes.

Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호길이(미터법)직경(미터법)입자 크기
30205-05463050 mm4.6 mm5 μm
30205-104630100 mm4.6 mm5 μm
30205-124630125 mm4.6 mm5 μm
30205-154630150 mm4.6 mm5 μm
30205-154030150 mm4 mm5 μm
30205-254030250 mm4 mm5 μm
30205-202130200 mm2.1 mm5 μm
30205-254630250 mm4.6 mm5 μm
30205-204630200 mm4.6 mm5 μm
30205-01400110mm4mm5µm
30205-253030250 mm3 mm5 μm
30210-204630200 mm4.6 mm10 μm
30210-254630250 mm4.6 mm10 μm
카탈로그 번호 30205-054630
제품 가격(KRW)
-
길이(미터법):
50 mm
직경(미터법):
4.6 mm
입자 크기:
5 μm

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable

Ordering Information:

Other column dimensions are available. Also available in 10 μm. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
사양
컬럼 형식Analytical Column
컬럼 유형Reversed Phase
최대 압력5800 psi (400 bar)
팩킹 물질Spherical, Fully Porous Type A Silica
정지상C8
표면적170 m²/g
온도60°C
USP 유형L7
탄소 부하6.5%
직경(미터법)4.6 mm
길이(미터법)50 mm
입자 크기5 μm
Reversed Phase
기공 크기120 Å
제품라인Hypersil Classical
pH2 to 8
Unit SizeEach