Search
Search
Use as a general-purpose C8 phase for acidic and neutral analytes.
| Catalog Number | Length (Metric) | Diameter (Metric) | Particle Size |
|---|---|---|---|
| 30205-154030 | 150 mm | 4 mm | 5 μm |
| 30205-054630 | 50 mm | 4.6 mm | 5 μm |
| 30203-124030 | 125 mm | 4 mm | 3 μm |
| 30205-104630 | 100 mm | 4.6 mm | 5 μm |
| 30205-102130 | 100 mm | 2.1 mm | 5 μm |
| 30205-124630 | 125 mm | 4.6 mm | 5 μm |
| 30205-154630 | 150 mm | 4.6 mm | 5 μm |
| 30205-254030 | 250 mm | 4 mm | 5 μm |
| 30205-254630 | 250 mm | 4.6 mm | 5 μm |
| 30205-204630 | 200 mm | 4.6 mm | 5 μm |
| 30205-014001 | 10 mm | 4 mm | 5 μm |
| 30205-253030 | 250 mm | 3 mm | 5 μm |
| 30210-204630 | 200 mm | 4.6 mm | 10 μm |
| 30210-254630 | 250 mm | 4.6 mm | 10 μm |
| 30203-054630 | 50 mm | 4.6 mm | 3 μm |
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.
Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
Ordering Information:
Other column dimensions are available. Also available in 10 μm. Please call Customer Service for more information.