2',4'-Dichloroacetophenone, 97%
2',4'-Dichloroacetophenone, 97%
2',4'-Dichloroacetophenone, 97%
Thermo Scientific Chemicals

2',4'-Dichloroacetophenone, 97%

CAS: 2234-16-4 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
Packaging:
Glass bottle
Catalog number 302430250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2234-16-4
IUPAC Name1-(2,4-dichlorophenyl)ethan-1-one
Molecular FormulaC8H6Cl2O
InChI KeyXMCRWEBERCXJCH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow-brown
GC>=96.0 %
Melting point29°C to 33°C
Infrared spectrumConforms
Appearance (Form)Solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only