Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns
Actual product may vary
Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns
Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns
Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns
Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS16-4μm Analytical, Capillary, and Guard IC Columns

Green features

Determine trace levels of perchlorate and other highly polarizable anions in drinking water, surface water, and ground water with these anion exchange columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Column Type
3027534 mmAnalytical Column
3027570.4 mmCapillary Column
3027580.4 mmGuard Column
3027552 mmAnalytical Column
3027544 mmGuard Column
302756TS2 mmGuard Column
Catalog number 302753
Price (MXN)
-
Diameter (Metric):
4 mm
Column Type:
Analytical Column

Determine trace levels of perchlorate and other highly polarizable anions in drinking water, surface water, and ground water with these anion exchange columns.

Designed for simple, sensitive analysis
  • Easily measure trace concentrations of perchlorate in drinking water, surface water, and ground water matrices using a large-loop injection
  • Improved efficiencies and resolution compared to the standard Dionex IonPac AS16 column
  • Easily transfer applications from the Dionex IonPac AS16 column to the Dionex IonPac AS16-4μm column due to their similar selectivity
  • Ultralow hydrophobicity allows for fast analysis of highly polarizable anions (thiosulfate, iodide, thiocyanate, and perchlorate) with a simple, isocratic eluent
  • Ideal for polyvalent anions, including polyphosphates
Use with a high-pressure IC system

Higher resolution is achieved through the use of smaller resin particles, which produce a column backpressure over 3000 psi at the standard flow rates. A high-pressure IC system, such as the Thermo Scientific™ Dionex™ ICS-5000+ HPIC™ system, Dionex Integrion™ HPIC system, or the Dionex ICS-4000 Integrated Capillary HPIC system is required to operate this column under standard conditions.

Reduce operating costs with lower eluent requirements

Capillary Dionex IonPac AS16-4μm columns provide the same performance as the standard bore and microbore Dionex IonPac AS16-4μm columns because they are packed with the same material. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system, requiring less eluent and producing less waste.

Multiple Formats Available

  • 4 x 250 mm and 2 x 250 mm analytical columns
  • 4 x 50 mm and 2 x 50 mm analytical guard columns
  • 0.4 x 250 mm capillary columns
  • 0.4 x 50 mm capillary guard columns
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation, and lower operating costs
Specifications
Anion Exchange Capacity170 μeq
Column Capacity170 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate1.5 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)Dionex IonPac AG16-4μm Standard Bore Guard Column
Green FeaturesFSC
HydrophobicityUltralow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure5000 psi (345 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size4 μm
QuantityEach
Solvent CompatibilitypH 0 to 14, 0 to 100% Organic HPLC Solvents
Stationary PhaseSuper Macroporous Resin
Column TypeAnalytical Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)250 mm
Unit SizeEach