L-Proline methyl ester hydrochloride, 98%
L-Proline methyl ester hydrochloride, 98%
L-Proline methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Proline methyl ester hydrochloride, 98%

CAS: 2133-40-6 | C6H12NO2 | 130.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 303000250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2133-40-6
IUPAC Name(2S)-2-(methoxycarbonyl)pyrrolidin-1-ium
Molecular FormulaC6H12NO2
InChI KeyBLWYXBNNBYXPPL-YFKPBYRVSA-O
SMILESCOC(=O)[C@@H]1CCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Argentometric>=97.5 %
Water=<0.5 % (K.F.)
Infrared spectrumConforms
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only