N-(tert-Butoxycarbonyl)ethanolamine, 98%
N-(tert-Butoxycarbonyl)ethanolamine, 98%
N-(tert-Butoxycarbonyl)ethanolamine, 98%
Thermo Scientific Chemicals

N-(tert-Butoxycarbonyl)ethanolamine, 98%

CAS: 26690-80-2 | C7H15NO3 | 161.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Catalog number 303170050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS26690-80-2
IUPAC Nametert-butyl N-(2-hydroxyethyl)carbamate
Molecular FormulaC7H15NO3
InChI KeyGPTXCAZYUMDUMN-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4475 to 1.4505 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only