2,5-Difluorobenzaldehyde, 98%
2,5-Difluorobenzaldehyde, 98%
2,5-Difluorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,5-Difluorobenzaldehyde, 98%

CAS: 2646-90-4 | C7H4F2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 303520250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2646-90-4
IUPAC Name2,5-difluorobenzaldehyde
Molecular FormulaC7H4F2O
InChI KeyVVVOJODFBWBNBI-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4970 to 1.5010 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only