3,5-Bis(trifluoromethyl)benzoyl chloride, 97%
3,5-Bis(trifluoromethyl)benzoyl chloride, 97%
3,5-Bis(trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzoyl chloride, 97%

CAS: 785-56-8 | C9H3ClF6O | 276.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3035300505 gGlass bottle
Catalog number 303530050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS785-56-8
IUPAC Name3,5-bis(trifluoromethyl)benzoyl chloride
Molecular FormulaC9H3ClF6O
InChI KeyWAKMMQSMEDJRRI-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(=C1)C(Cl)=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4322 to 1.4342 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only