alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
Thermo Scientific Chemicals

alpha-Bromo-2,6-difluorotoluene, 97%

CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 303570250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS85118-00-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point51°C to 55°C
Infrared spectrumConforms
Appearance (Form)Crystals and/or chunks
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only