3,5-Bis(trifluoromethyl)benzyl bromide, 97%
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzyl bromide, 97%

CAS: 32247-96-4 | C9H5BrF6 | 307.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 303640010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS32247-96-4
IUPAC Name1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
Molecular FormulaC9H5BrF6
InChI KeyATLQGZVLWOURFU-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only