2,2,3,3,3-Pentafluoro-1-propanol, 97%
2,2,3,3,3-Pentafluoro-1-propanol, 97%
2,2,3,3,3-Pentafluoro-1-propanol, 97%
Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoro-1-propanol, 97%

CAS: 422-05-9 | C3H3F5O | 150.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 303790050
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.20
Save 5.55
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.20
Save 5.55
Each
Chemical Identifiers
CAS422-05-9
IUPAC Name2,2,3,3,3-pentafluoropropan-1-ol
Molecular FormulaC3H3F5O
InChI KeyPSQZJKGXDGNDFP-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only