3-Methyl-1-cyclohexene, 90%, Tech.
3-Methyl-1-cyclohexene, 90%, Tech.
3-Methyl-1-cyclohexene, 90%, Tech.
Thermo Scientific Chemicals

3-Methyl-1-cyclohexene, 90%, Tech.

CAS: 591-48-0 | C7H12 | 96.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
30431025025 mLGlass bottle
Catalog number 304310250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-48-0
IUPAC Name3-methylcyclohex-1-ene
Molecular FormulaC7H12
InChI KeyUZPWKTCMUADILM-UHFFFAOYNA-N
SMILESCC1CCCC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=88 %
Appearance (Form)Liquid
Refractive index1.4425 to 1.4445 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only