Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 lb.
Número de catálogo 3045-05
Precio (MXN)
-
Cantidad:
2.5 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorBlanco
Punto de fusión773°C
EnvasePoly bottle, tamper-evident cap
Punto de ebullición1420°C (1013 hPa)
GradoMulticomponente/USP
Peso molecular (g/mol)74.55
Porcentaje de pureza99 to 105%
Forma físicaSolid
Cantidad2.5 kg
pH7 (15°C, saturated solution in water)
Unit SizeEach