Dimethyl bromomalonate, 90%, tech.
Dimethyl bromomalonate, 90%, tech.
Dimethyl bromomalonate, 90%, tech.
Thermo Scientific Chemicals

Dimethyl bromomalonate, 90%, tech.

CAS: 868-26-8 | C5H7BrO4 | 211.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
30519025025 mLGlass bottle
Catalog number 305190250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS868-26-8
IUPAC Name1,3-dimethyl 2-bromopropanedioate
Molecular FormulaC5H7BrO4
InChI KeyNEMOJKROKMMQBQ-UHFFFAOYSA-N
SMILESCOC(=O)C(Br)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=88 %
Infrared spectrumConforms
Refractive index1.4580 to 1.4660 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only