3,4-Dimethoxybenzoyl chloride, 98%
3,4-Dimethoxybenzoyl chloride, 98%
3,4-Dimethoxybenzoyl chloride, 98%
Thermo Scientific Chemicals

3,4-Dimethoxybenzoyl chloride, 98%

CAS: 3535-37-3 | C9H9ClO3 | 200.62 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 305600050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3535-37-3
IUPAC Name3,4-dimethoxybenzoyl chloride
Molecular FormulaC9H9ClO3
InChI KeyVIOBGCWEHLRBEP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Powder or crystals or flakes and/or chunks
Infrared spectrumConforms
Appearance (Color)White to yellow to grey
Melting point68°C to 74°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only