1,1-Bis(methylthio)-2-nitroethylene, 99%
1,1-Bis(methylthio)-2-nitroethylene, 99%
1,1-Bis(methylthio)-2-nitroethylene, 99%
Thermo Scientific Chemicals

1,1-Bis(methylthio)-2-nitroethylene, 99%

CAS: 13623-94-4 | C4H7NO2S2 | 165.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 305690250
Price (EUR)
178,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
178,00
Each
Chemical Identifiers
CAS13623-94-4
IUPAC Name1,1-bis(methylsulfanyl)-2-nitroethene
Molecular FormulaC4H7NO2S2
InChI KeyNXGHEDHQXXXTTP-UHFFFAOYSA-N
SMILESCSC(SC)=C[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting point124°C to 127°C
Appearance (Form)Powder
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only