Thermo Scientific™
Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope Slides
 Related applications:
Anatomical Pathology
Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass. Choose from sandblast frosted end or white epoxy frosted end for easy marking. Each tab is impervious to virtually all common laboratory chemicals and reagents.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.
  • Smooth marking area for effortless writing, automated printing, or printed labels
  • Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
  • Slide boxes convert into convenient slide storage files
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Part No. 3032-002 is used for fluorescent antibody examination
Documents
Manuals & protocols