Etil 6-bromohexanoato, 98 %, Thermo Scientific Chemicals
Etil 6-bromohexanoato, 98 %, Thermo Scientific Chemicals
Etil 6-bromohexanoato, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Etil 6-bromohexanoato, 98 %, Thermo Scientific Chemicals

CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 305920250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS25542-62-5
IUPAC Nameethyl 6-bromohexanoate
Molecular FormulaC8H15BrO2
InChI KeyDXBULVYHTICWKT-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCCBr
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4580 to 1.4600 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only