4-(Trifluoromethoxy)phenol, 98%
4-(Trifluoromethoxy)phenol, 98%
4-(Trifluoromethoxy)phenol, 98%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenol, 98%

CAS: 828-27-3 | C7H5F3O2 | 178.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 306040010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS828-27-3
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4440 to 1.4490 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only