Methyl vinyl sulfone, 95%, stabilized
Methyl vinyl sulfone, 95%, stabilized
Methyl vinyl sulfone, 95%, stabilized
Thermo Scientific Chemicals

Methyl vinyl sulfone, 95%, stabilized

CAS: 3680-02-2 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 306730010
Price (EUR)
30,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,10
Each
Chemical Identifiers
CAS3680-02-2
IUPAC Namemethanesulfonylethene
Molecular FormulaC3H6O2S
InChI KeyWUIJTQZXUURFQU-UHFFFAOYSA-N
SMILESCS(=O)(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Stabilizer200 ppm 4-tert-Butylphenol
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4610 to 1.4650 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only