Ethyl ethynyl ether, 50 wt% solution in hexanes
Ethyl ethynyl ether, 50 wt% solution in hexanes
Ethyl ethynyl ether, 50 wt% solution in hexanes
Thermo Scientific Chemicals

Ethyl ethynyl ether, 50 wt% solution in hexanes

CAS: 927-80-0 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 307390250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS927-80-0
IUPAC Nameethoxyethyne
Molecular FormulaC4H6O
InChI KeyWMYNMYVRWWCRPS-UHFFFAOYSA-N
SMILESCCOC#C
View more
SpecificationsSpecification SheetSpecification Sheet
GC45 to 55 % w/w
Appearance (Color)Clear colorless to yellow to red to brown
Appearance (Form)Solution
Refractive index1.3750 to 1.3850 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only