3,5-Bis(trifluoromethyl)benzaldehyde, 97%
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
3,5-Bis(trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzaldehyde, 97%

CAS: 401-95-6 | C9H4F6O | 242.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 307690250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS401-95-6
IUPAC Name3,5-bis(trifluoromethyl)benzaldehyde
Molecular FormulaC9H4F6O
InChI KeyLDWLIXZSDPXYDR-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(C=O)=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4215 to 1.4235 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only