cis-2,3-Epoxybutane, 97%
cis-2,3-Epoxybutane, 97%
cis-2,3-Epoxybutane, 97%
Thermo Scientific Chemicals

cis-2,3-Epoxybutane, 97%

CAS: 1758-33-4 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 307870050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1758-33-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3820 to 1.3840 (20°C, 589 nm)
GC>=96.0 % (corrected for H2O)
Water=<2.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only