Regulatory requirements vary by country; products may not be available in your geographic area.
PrioSTRIP™ Reading Device V700
Catalog number: 30800
 Related applications: Animal Health
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Scanner with frame in which PrioSTRIP™ combs can be inserted. Used for the evaluation of Prionics®-Check PrioSTRIP test results.
For Veterinary Use Only.
Specifications
For Use With (Equipment): 30000 PrioSTRIP™
Product Line: PrioSTRIP™, Prionics™
Product Size: 1 piece
Type: Scanner
Documents
Manuals & protocols