Tin(II) fluoride, 99%
Tin(II) fluoride, 99%
Tin(II) fluoride, 99%
Thermo Scientific Chemicals

Tin(II) fluoride, 99%

CAS: 7783-47-3 | F2Sn | 156.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 308501000
Price (USD)
87.70
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
87.70
Each
Chemical Identifiers
CAS7783-47-3
IUPAC Nameλ²-tin(2+) difluoride
Molecular FormulaF2Sn
InChI KeyANOBYBYXJXCGBS-UHFFFAOYSA-L
SMILES[F-].[F-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Titration Iodimetric>=98.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only