4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals

CAS: 943-27-1 | Molecular Formula: C12H16O | Molecular Weight: 176.26 g/mol
Have Questions?
Catalog NumberQuantity
3087400505 g
Catalog number 308740050
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.10
Save 14.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.10
Save 14.45
Each
Chemical Identifiers
CAS943-27-1
IUPAC Name1-(4-tert-butylphenyl)ethan-1-one
Molecular FormulaC12H16O
InChI KeyUYFJYGWNYQCHOB-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5190 to 1.5220 (20°C, 589 nm)
GC>=97.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only