Tetraethyl methylenediphosphonate, 97%
Tetraethyl methylenediphosphonate, 97%
Tetraethyl methylenediphosphonate, 97%
Tetraethyl methylenediphosphonate, 97%
Thermo Scientific Chemicals

Tetraethyl methylenediphosphonate, 97%

CAS: 1660-94-2 | C9H22O6P2 | 288.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 308820250
Price (EUR)
255,00
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
255,00
Each
Add to cart
Chemical Identifiers
CAS1660-94-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4400 to 1.4440 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only