2-Tolylboronic acid, 95%
2-Tolylboronic acid, 95%
2-Tolylboronic acid, 95%
Thermo Scientific Chemicals

2-Tolylboronic acid, 95%

CAS: 16419-60-6 | C7H9BO2 | 135.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 309260050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16419-60-6
IUPAC Name(2-methylphenyl)boronic acid
Molecular FormulaC7H9BO2
InChI KeyNSJVYHOPHZMZPN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
HPLC>=94 %
Infrared spectrumConforms
Appearance (Color)White to beige
Melting point160°C to 166°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only