1,6-Heptadiyne, 98%
1,6-Heptadiyne, 98%
1,6-Heptadiyne, 98%
Thermo Scientific Chemicals

1,6-Heptadiyne, 98%

CAS: 2396-63-6 | C7H8 | 92.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 309420010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2396-63-6
IUPAC Namehepta-1,6-diyne
Molecular FormulaC7H8
InChI KeyRSPZSDWVQWRAEF-UHFFFAOYSA-N
SMILESC#CCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4400 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only