L-Aspartyl-L-phenylalanine, 95%
L-Aspartyl-L-phenylalanine, 95%
L-Aspartyl-L-phenylalanine, 95%
Thermo Scientific Chemicals

L-Aspartyl-L-phenylalanine, 95%

CAS: 13433-09-5 | C13H16N2O5 | 280.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
Catalog number 310021000
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.20
Save 12.55
Each
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.20
Save 12.55
Each
Chemical Identifiers
CAS13433-09-5
IUPAC Name(3S)-3-amino-3-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propanoic acid
Molecular FormulaC13H16N2O5
InChI KeyYZQCXOFQZKCETR-UWVGGRQHSA-N
SMILESN[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+10.0° to +14.0° (20°C, 589 nm) (c=1, 0.5N HCl)
Appearance (Form)Powder
HPLC>=94 %
Appearance (Color)White to almost white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only