ART™ Reload System
Thermo Scientific™
ART™ Reload System
Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)For Use With (Equipment)Tip StyleFit for PurposeTamanho da unidade
3102-05-RT1000 μLUniversal FitLow RetentionCertified QualityCase of 2800
3101-05-RT1000 μLUniversal FitLow RetentionSterileCase of 2800
3101-05-RTPK
conhecido também como 3101-05-RT
1000 μLUniversal FitLow RetentionSterilePack of 560
3522-RTPK
conhecido também como 3522-RT
20 μLEppendorfStandardCertified QualityPack of 960
3101-RT1000 μLUniversal FitStandardSterileCase of 2800
3102-05-RTPK
conhecido também como 3102-05-RT
1000 μLUniversal FitLow RetentionCertified QualityPack of 560
3501-05-RT10 μLUniversal FitLow RetentionSterileCase of 4800
3501-RT10 μLUniversal FitStandardSterileCase of 4800
3501-RTPK10 μLUniversal FitStandardSterilePack of 960
3502-05-RT10 μLUniversal FitLow RetentionCertified QualityCase of 4800
3502-05-RTPK
conhecido também como 3502-05-RT
10 μLUniversal FitLow RetentionCertified QualityPack of 960
3502-RT10 μLUniversal FitStandardCertified QualityCase of 4800
3521-05-RT20 μLEppendorfLow RetentionSterileCase of 4800
3521-RT20 μLEppendorfStandardSterileCase of 4800
3521-RTPK
conhecido também como 3521-RT
20 μLEppendorfStandardSterilePack of 960
3522-05-RT20 μLEppendorfLow RetentionCertified QualityCase of 4800
3522-05-RTPK
conhecido também como 3522-05-RT
20 μLEppendorfLow RetentionCertified QualityPack of 960
3522-RT20 μLEppendorfStandardCertified QualityCase of 4800
3551-05-RT200 μLUniversal FitLow RetentionSterileCase of 4800
3551-RT200 μLUniversal FitStandardSterileCase of 4800
3552-05-RT200 μLUniversal FitLow RetentionCertified QualityCase of 4800
3552-RT200 μLUniversal FitStandardCertified QualityCase of 4800
3771-05-RT300 μLUniversal FitLow RetentionSterileCase of 4800
3771-05-RTPK
conhecido também como 3771-05-RT
300 μLUniversal FitLow RetentionSterilePack of 960
3771-RT300 μLUniversal FitStandardSterileCase of 4800
3771-RTPK
conhecido também como 3771-RT
300 μLUniversal FitStandardSterilePack of 960
3772-05-RT300 μLUniversal FitLow RetentionCertified QualityCase of 4800
3772-05-RTPK
conhecido também como 3772-05-RT
300 μLUniversal FitLow RetentionCertified QualityPack of 960
3772-RT300 μLUniversal FitStandardCertified QualityCase of 4800
3772-RTPK
conhecido também como 3772-RT
300 μLUniversal FitStandardCertified QualityPack of 960
Número do catálogo 3102-05-RT
Preço (BRL)
-
Volume (Metric):
1000 μL
For Use With (Equipment):
Universal Fit
Tip Style:
Low Retention
Fit for Purpose:
Certified Quality
Unit Size:
Case of 2800
Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.
Features:
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Note:
Empty rack required for -RT parts
Especificações
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
Length (English)3.5 in.
Length (Metric)8.89 cm
ModelART Non-filtered
Quantity2800/Cs.
ColorNatural
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilityNon-sterile
Tip StyleLow Retention
Volume (Metric)1000 μL
Unit SizeCase of 2800