HLA-DR Rabbit Complement
Catalog number: 31023100
 Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
8-12 Week Complemement
• Produced from large pools of New Zealand and California White (albino) rabbits 8-12 weeks of age.
• Collected under controlled methods to ensure the integrity and quality of the complement.
• Application: For use in HLA lymphocytotoxic assays.
• Tested for high titer and low background toxicity.
3-4 Week Complement
• The low heterophile activity in 3-4 week complement results in low background, making this product ideal for many uses within the research laboratory, including bactericidal and opsonization assays.
Contents & storage
Store in ultra-cold freezer (-68 to -85°C).
Documents
Manuals & protocols