Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals

Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 310240100
Price (JPY)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range90%
CAS676-83-5
Chemical Name or MaterialDichloromethylphosphine
Density1.3000g/mL
Formula Weight116.92
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only