Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 310240500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS676-83-5
IUPAC Namedichloro(methyl)phosphane
Molecular FormulaCH3Cl2P
InChI KeyCDPKWOKGVUHZFR-UHFFFAOYSA-N
SMILESCP(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=88 %
Refractive index1.4945 to 1.4985 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only