Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Thermo Scientific™

Nalgene™ PPCO Conical-Bottom Centrifuge Tubes

Simplify measurements using these PPCO conical-bottom centrifuge tubes, which feature molded-in graduations. Translucent PPCO has excellent chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)Unit Size
3103-0050PK
also known as 3103-0050
50 mL133.1 mm28.7 mmPack of 10
3103-001515 mL118.6 mm16.8 mmCase of 100
3103-0015PK
also known as 3103-0015
15 mL118.6 mm16.8 mmPack of 10
3103-005050 mL133.1 mm28.7 mmCase of 100
Catalog number 3103-0050PK
also known as 3103-0050
Price (USD)/ Pack of 10
54.97
Online Exclusive
123.30
Save 68.33 (55%)
Add to cart
Capacity (Metric):
50 mL
Length (Metric):
133.1 mm
Diameter (Metric):
28.7 mm
Unit Size:
Pack of 10
Price (USD)/ Pack of 10
54.97
Online Exclusive
123.30
Save 68.33 (55%)
Add to cart

Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.

Features:
  • Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
  • Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)
  • Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
  • Molded-in graduations last the life of the tube
  • Last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Tube are both autoclavable and graduated
  • Autoclavable

Ordering Information:

Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.

Recycling Information: 
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.

Specifications
Diameter (Metric)28.7 mm
GraduationsMolded-in
No. per Case100
No. per Pack10
AutoclavableAutoclavable
Bottom ShapeConical
Capacity (Metric)50 mL
Length (Metric)133.1 mm
MaterialPolypropylene Copolymer
Max. RCF6000 x g
Product TypeCentrifuge Tube
ShapeConical
SterilityNon-sterile
Writing AreaNo
Unit SizePack of 10