Search
View additional product information for AIM V™ Medium (research grade), AlbuMAX™ Supplement - FAQs (31035025)
1 product FAQs found
There is no extra supplementation required for this product, but we do recommend refreshing L-glutamine supplementation every 30 days.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.