Forma™ Series II 3110 Water-Jacketed CO2 Incubator, 184 L, Polished Stainless Steel - FAQs
Forma™ Series II 3110 Water-Jacketed CO2 Incubator, 184 L, Polished Stainless Steel - FAQs
View additional product information for Forma™ Series II Water-Jacketed CO2 Incubator, 184L - FAQs (3111, 3110, 3121, 3131, 3120, 3130, 3141, 3140)
1 product FAQs found
What amperage does the Forma Series II Water-Jacketed CO2 Incubator, 184L (Cat. No. 3110) draw?
The Forma Series II Water-Jacketed CO2 Incubator, 184L (Cat. No. 3110) draws 3.6 FLA (Full Load Amps) at 115V, 50/60 Hz.