Pierce™ Immunodiffusion Plates, Multiple Pattern
Thermo Scientific™

Pierce™ Immunodiffusion Plates, Multiple Pattern

Thermo Scientific Pierce Immunodiffusion Plates are standard Ouchterlony gel plates containing patterns of six wells around one well for studyingRead more
Have Questions?
Catalog NumberQuantity
311116 plates
Catalog number 31111
Price (USD)
236.00
Each
Quantity:
6 plates
Price (USD)
236.00
Each
Thermo Scientific Pierce Immunodiffusion Plates are standard Ouchterlony gel plates containing patterns of six wells around one well for studying and characterizing antibody-antigen binding interactions.

Features of Pierce Immunodiffusion Plates:

• Gelling agent contains precipitin brighteners for easy visualization
• Immunodiffusion plates contain diffusion enhancers to help speed the interaction process
• Excellent precipitin bands with antibodies from all species (including rabbit antibodies)
• Gels can be washed, dried and stained for a permanent record

Immunodiffusion (ID) is a classic technique for detecting antibody-antigen interactions based on the theory of double diffusion originally described by Oudin and Ouchterlony. Antigens and antibodies are placed into separate wells that are cut into a gel matrix and allowed to diffuse towards each other. If the reaction is positive, a precipitate forms that appears as an opaque line. The precipitation reaction occurs when the antigen and antibody concentrations are combined at near equivalent proportions.

When multivalent antigens combine with divalent antibodies in solution, three-dimensional lattices are formed that aggregate and precipitate. The amount of precipitate varies in proportion to the concentration of the antigen and antibody. At equivalent or optimal proportions almost all the antigen and antibody will precipitate. If there is an excess of antibody, the complexes formed with the antigen are insoluble. When there is an excess of antigen, the precipitate has a tendency to dissolve caused by the formation of soluble complexes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayImmunodiffusion (ID)
Detection MethodPrecipitate
Product LinePierce
Quantity6 plates
Target SpecificityNot Target-Specific
TechniqueImmunodiffusion (ID)
TypeMulti-Well Plate
Volume (Metric) Working22 μL
ColorClear
Diameter (Metric)3.2 mm
Height (Metric)2.8 mm
Well Count6
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Plates are shipped at ambient temperature. Do not freeze plates or allow them to become dry.

Frequently asked questions (FAQs)

What do I need to consider when designing a peptide for antibody production?

We recommend that hydrophobic residues comprise 50% or less of all the residues in your sequence. Make sure there is at least one charged residue for every five amino acids: this is generally known to enhance the solubility of the peptide. Peptides (compared to polypeptides, which fold and bury the hydrophobic amino acids) are too small to fold, so just a few hydrophobic amino acids may leave them insoluble.

Other guidelines include:
Peptides containing multiple Cys, Met, and Trp can be hard to synthesize.
Some sequences are problematic in solid-phase peptide synthesis or cleavage and are best avoided; these include Asp-Pro sequences or stretches of amino acids that require bulky protecting groups on their side chains during synthesis.
Glycine is often good for antigenicity as it has only a hydrogen side chain; this allows for complete rotation.
Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What is the optimal peptide length for antibody production?

Most peptide antigens range in length from 12 to 16 residues and are relatively easy to synthesize. Peptides of 9 residues or shorter have been effective antigens for antibody production, but peptides longer than 16 amino acids may contain several epitopes and form secondary structures. Peptides in excess of 18 residues begin to present more synthetic challenges. Before you synthesize your peptide, we recommend doing a BLASTP search using your peptide sequence. This is to make sure that the peptide is not homologous or identical to a sequence in a completely unrelated protein in the host animal.

Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What happens if antibodies are stored improperly?

Improper storage of antibodies can lead to:
- Degradation: Loss of activity and specificity
- Aggregation: Formation of precipitates or clumps
- Reduced performance: Poor results in assays or experiments

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

What do antibody pair kits contain, how many samples can I process, and how can I find a list of antibody pair kits by target?

Antibody pair kits contain capture antibody, detection antibody, recombinant standard and HRP conjugate. Each contains enough reagents to process forty 96-well plates. A list of Antibody Pair Kits (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/elisa/antibody-pair-kits.html) is available by target.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

What do Reagent Sets contain, how many samples can I process, and how can I check if a Reagent Set is available for my target?

Reagent Sets contain Capture Antibody, Detection Antibody, Recombinant Standard, HRP Conjugate, TMB Substrate and Stop Solution. Each contains enough reagents to process five 96-well plates. Reagent Sets are included in the main list of ELISA (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/elisa/antibody-pair-kits.html) kits (search by “Reagent Set”).

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.