Thermo Scientific™
VisionMate™ SR Single Rack 2D Barcode Reader
 Related applications: Lab Automation
Decode a full rack of barcoded storage tubes in less than ten seconds with the Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader. This compact, user-friendly reader provides exceptional flexibility with data export formats, tube type readability, and tube layout options.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Controlled via on-screen software interface or remotely via TCP/IP connection
  • Scans a full tube rack in less than ten seconds
  • Simply follow the instructions during software installation and connect the reader to a computer via USB port
  • Integrates easily with LIMS, existing databases or tracking systems
  • Small footprint and compact profile save valuable laboratory space
Download software updates
Visit our Sample Storage Software Downloads page to update your software for the VisionMate High Speed Barcode Reader and other sample storage equipment.
Documents
Manuals & protocols